Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Imibenconazole Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018494 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8494 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8494 Supplier MilliporeSigma Supelco Supplier No. 327252ML

Imibenconazoles belong to the family of azole-type fungicides.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 86598-92-7
Flash Point 4.0°C - Closed Cup
Linear Formula C17H13 Cl3N4 S
MDL Number MFCD01076617
Synonym 4-Chlorobenzyl N-(2,4-dichlorophenyl)-1 H-1,2,4-triazole-1-acetimidothioate
Type Analytical Standard
Quantity 2 mL
Formula Weight 411.74
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.