Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Imidocarb dipropionate, MilliporeSigma™ Supelco™

Catalog No. 111014610 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4610 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4610 Supplier MilliporeSigma Supelco Supplier No. 3344150MG
Add to Cart
Add to Cart

Imidocarb dipropionate is an antibiotic and diaminide of the carbanilide series of antiprotozoal compounds. It is commonly used in animals like horses, where it is found to be effective in inhibiting the Theileria equi strains causing infections.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 55750-06-6
Physical Form Neat
Linear Formula C25H32N6O5
MDL Number MFCD00013161
Melting Point 205°C
Synonym N,N′-Bis[3-(4,5-dihydro-1 H-imidazol-2-yl)phenyl]urea dipropanoate
Type Analytical Standard
Quantity 50 mg
Formula Weight 496.56
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.