Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Ipronidazole-OH-(N-methyl-d3), MilliporeSigma™ Supelco™

Catalog No. 111012649 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2649 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2649 Supplier MilliporeSigma Supelco Supplier No. 3218910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Line VETRANAL
CAS 1156508-86-9
Physical Form Neat
Linear Formula C7D3H8N3O3
MDL Number MFCD26142912
Synonym 2-[1-(Methyl-d3)-5-nitro-1 H-imidazol-2-yl]propan-2-ol; Hydroxy-ipronidazole-d3; IPZOH-d3
Type Analytical Standard
Quantity 10 mg
Formula Weight 188.20
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.