Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Levamisol hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111019519 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9519 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9519 Supplier MilliporeSigma Supelco Supplier No. 31742250MG
Add to Cart
Add to Cart

Levamisol hydrochloride is an anthelmintic drug which possesses immunomodulatory and anticancer properties. It is a white crystalline compound which is stable in acid aqueous media, whereas it hydrolyzes in alkaline/neutral solutions.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 16595-80-5
Physical Form Neat
Linear Formula C11H12N2 S · HCl
MDL Number MFCD00012675
Melting Point 228°C to 230°C
Synonym (-)-Tetramisole hydrochloride; (S)-(-)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride; L(-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride; Levamisole hydrochloride
Type Analytical Standard
Quantity 250 mg
Formula Weight 240.75
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.