Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Lincomycin hydrochloride monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111015340 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5340 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5340 Supplier MilliporeSigma Supelco Supplier No. 31727250MG
Add to Cart
Add to Cart

Chemical structure: aminoglycoside

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 7179-49-9
Physical Form Neat
Linear Formula C18H34N2O6 S · HCl · H2O
MDL Number MFCD00167245
Type Analytical Standard
Quantity 250 mg
Formula Weight 461.01
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.