Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Loperamide hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111020167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0167 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0167 Supplier MilliporeSigma Supelco Supplier No. 34014100MGR
Add to Cart
Add to Cart

Loperamide is an antidiarrheal agent used in controlling acute nonspecific diarrhea and chronic diarrhea associated with inflammatory bowel diseases.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 34552-83-5
Physical Form Neat
Linear Formula C29H33 ClN2O2 · HCl
MDL Number MFCD00058581
Synonym 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutyramide hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 513.50
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.