Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Maduramicin ammonium, MilliporeSigma™ Supelco™

Catalog No. 111019184 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9184 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9184 Supplier MilliporeSigma Supelco Supplier No. 34069100MG
Add to Cart
Add to Cart

Maduramicin ammonium belongs to the class of antibiotics employed as veterinary drugs to improve animal health and promote growth.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 84878-61-5
Physical Form Neat
Linear Formula C47H83 NO17
MDL Number MFCD00865282
Synonym Maduramycin ammonium
Type Analytical Standard
Quantity 100 mg
Formula Weight 934.16
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.