Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Melengestrol acetate, MilliporeSigma™ Supelco™

Catalog No. 111013835 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3835 Supplier MilliporeSigma Supelco Supplier No. 33998100MGR
Add to Cart
Add to Cart

Melengestrol acetate belongs to the family of hormones. This acetylgestagen compound is known to elicit a gestagenic effect in vivo.

TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 2919-66-6
Physical Form Neat
Linear Formula C25H32O4
MDL Number MFCD00072142
Melting Point 202°C to 204°C
Synonym 17 alpha-Acetoxy-6-methyl-16-methylene-4,6-pregnadiene-3,20-dione; MGA
Type Analytical Standard
Quantity 100 mg
Formula Weight 396.52
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.