Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Neomycin trisulfate salt hydrate, MilliporeSigma™ Supelco™

Catalog No. 111013873 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3873 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3873 Supplier MilliporeSigma Supelco Supplier No. 33492100MGR
Add to Cart
Add to Cart

Chemical structure: aminoglycoside

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 1405-10-3
Physical Form Neat
Linear Formula C23H46N6O13 · 3H2 SO4 · xH2 O
Type Analytical Standard
Quantity 100 mg
Formula Weight 908.88 (anhydrous basis)
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.