Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Nitroxinil, MilliporeSigma™ Supelco™

Catalog No. 111018600 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8600 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8600 Supplier MilliporeSigma Supelco Supplier No. 34088100MG
Add to Cart
Add to Cart

Nitroxinil is an anthelmintic agent, which is also an antiparasite. It is highly effective against Fasciola species and its mode of action involves uncoupling the oxidative phosphorylation in liver flukes and also reduces spermatogenesis in surviving flukes, resulting in fewer fertile eggs.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 1689-89-0
Physical Form Neat
Linear Formula C7H3 IN2O3
MDL Number MFCD00070776
Synonym 4-Hydroxy-3-iodo-5-nitrobenzonitrile; Nitroxynil
Type Analytical Standard
Quantity 100 mg
Formula Weight 290.01
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.