Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Ractopamine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111015852 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5852 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5852 Supplier MilliporeSigma Supelco Supplier No. 34198100MG
Only null left
Add to Cart
Add to Cart

Ractopamine hydrochloride is a phenethanolamine with beta-adrenergic agonist properties, which can be used as animal feed additive for swine in order to increase growth performance.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 90274-24-1
Physical Form Neat
Linear Formula C18H23 NO3 · HCl
MDL Number MFCD07781271
Synonym 4-{3-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]aminobutylphenol hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 337.84
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.