Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ (S)-(-)-Sulpiride, MilliporeSigma™ Supelco™

Catalog No. 111013639 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3639 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3639 Supplier MilliporeSigma Supelco Supplier No. 34002100MGR
Add to Cart
Add to Cart

(S)-(−)-Sulpiride is a levorotatory enantiomer of sulpiride, which shows greater central antidopaminergic activity, antiemetic and antidyspeptic effects. It is also used as a therapeutic drug for the treatment of depression and somatoform disorders.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 23672-07-3
Physical Form Neat
Linear Formula C15H23N3O4 S
MDL Number MFCD00055220
Melting Point 183°C to 186°C
Synonym (S)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide; Levosulpiride
Type Analytical Standard
Quantity 100 mg
Formula Weight 341.43
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.