Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Sulfaclozine sodium, MilliporeSigma™ Supelco™

Catalog No. 111013616 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3616 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3616 Supplier MilliporeSigma Supelco Supplier No. 32421100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfaclozine has been used in an experimental study conducted to evaluate different persulfate activation methods.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 23307-72-4
Physical Form Neat
Linear Formula C10H8 ClN4 NaO2 S
MDL Number MFCD09954636
Type Analytical Standard
Quantity 100 mg
Formula Weight 306.70
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.