Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Sulfadiazine, MilliporeSigma™ Supelco™

Catalog No. 111015162 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5162 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5162 Supplier MilliporeSigma Supelco Supplier No. 35033100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical structure: sulfonamide

Environmental and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 68-35-9
Physical Form Neat
Linear Formula C10H10N4O2 S
MDL Number MFCD00006065
Melting Point 253°C
Synonym 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide; N1-(Pyrimidin-2-yl)sulfanilamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 250.28
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.