Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Sulfadimethoxine, MilliporeSigma™ Supelco™

Catalog No. 111012796 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2796 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2796 Supplier MilliporeSigma Supelco Supplier No. 46794250MG
Add to Cart
Add to Cart

Sulfadimethoxine is an long acting sulfonamide antibiotic, which finds applications in pen-held lobsters, Homarus americanus for treatment of gaffkemia.

Environmental and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 122-11-2
Physical Form Neat
Linear Formula C12H14N4O4 S
MDL Number MFCD00057345
Synonym 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 310.33
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.