Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Sulfamethoxazole-(phenyl-13C6), MilliporeSigma™ Supelco™

Catalog No. 111012734 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2734 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2734 Supplier MilliporeSigma Supelco Supplier No. 3251410MG
Add to Cart
Add to Cart

Chemical structure: sulfonamide

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 1196157-90-0
Physical Form Neat
Linear Formula 13C6C4H11N3O3 S
MDL Number MFCD16652518
Synonym 4-Amino-N-(5-methyl-3-isoxazolyl)benzene-13C6-sulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide-13C6
Type Analytical Standard
Quantity 10 mg
Formula Weight 259.23
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.