Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Sulfapyridine, MilliporeSigma™ Supelco™

Catalog No. 111013127 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3127 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3127 Supplier MilliporeSigma Supelco Supplier No. 31738250MG
Add to Cart
Add to Cart

Chemical structure: sulfonamide

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 144-83-2
Physical Form Neat
Linear Formula C11H11N3O2 S
MDL Number MFCD00038036
Synonym 4-Amino-N-[2-pyridyl]benzene sulfonamide; N1-(Pyridin-2-yl)sulfanilamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 249.29
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.