Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Tetracycline hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111020120 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0120 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0120 Supplier MilliporeSigma Supelco Supplier No. 31741250MG
Add to Cart
Add to Cart

Tetracycline hydrochloride (TCH) is an antibiotic drug for treating infections developed by gram-positive microorganism and anthrax infections. It also helps as an alternate drug for penicillin.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 64-75-5
Physical Form Neat
Linear Formula C22H24N2O8 · HCl
MDL Number MFCD00078142
Melting Point 220°C to 223°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 480.90
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.