Learn More
VETRANAL™ Tilmicosin, Analytical Standard, Mix of Isomers, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Product Line | VETRANAL |
| Physical Form | Neat |
| Linear Formula | C46H80N2O13 |
| MDL Number | MFCD19705582 |
| Synonym | 20-Deoxo-20-(3,5-dimethyl-1-piperidinyl)desmycosin |
| Quantity | 100 mg |
| Formula Weight | 869.13 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.