Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Triclabendazole sulfoxide, MilliporeSigma™ Supelco™

Catalog No. 111012278 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2278 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2278 Supplier MilliporeSigma Supelco Supplier No. 3539110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triclabendazole sulfoxide belongs to the benzimidazole class of anthelmintics drugs. It is very much effective against mature and immature stages of Fasciola hepatica and Fasciola gigantica in all ruminant species.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 100648-13-3
Physical Form Neat
Linear Formula C14H9 Cl3N2O2 S
MDL Number MFCD09038493
Synonym 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1 H-benzimidazole
Type Analytical Standard
Quantity 10 mg
Formula Weight 375.66
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.