Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Triflupromazine Hydrochloride Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022534 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2534 10 mg
11-102-2535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2534 Supplier MilliporeSigma Supelco Supplier No. 4697610MG

Specifications

CAS 1098-60-8
Molecular Formula C18H19F3N2S · HCl
Linear Formula C18H19F3N2S · HCl
MDL Number MFCD00058103
Quantity 10 mg
UN Number UN 2811 6.1/PGIII
Synonym N,N-Dimethyl-2-(trifluoromethyl)-10 H-phenothiazine-10-propanamine monohydrochloride
Formula Weight 388.88
Grade Analytical Standard
Physical Form Neat

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.