Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Tulobuterol hydrochloride, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014653 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4653 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4653 Supplier MilliporeSigma Supelco Supplier No. 5354110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tulobuterol hydrochloride is a sympathomimetic amine, possessing bronchodilator activity.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56776-01-3
Product Line VETRANAL
Physical Form Neat
Linear Formula C12H18 ClNO · HCl
MDL Number MFCD00214398
Synonym 2-tert-Butylamino-1-(2-chlorophenyl)ethanol hydrochloride
Type Analytical Standard
Quantity 10 mg
Formula Weight 264.19
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.