Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Valnemulin hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111012966 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2966 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2966 Supplier MilliporeSigma Supelco Supplier No. 3237525MG
Add to Cart
Add to Cart

Specifications

Product Line VETRANAL
CAS 133868-46-9
Physical Form Neat
Linear Formula C31H52N2O5 S · HCl
MDL Number MFCD08458958
Type Analytical Standard
Quantity 25 mg
Formula Weight 601.28
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.