Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Voriconazole, MilliporeSigma™ Supelco™

Catalog No. 111018896 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8896 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8896 Supplier MilliporeSigma Supelco Supplier No. 3248325MG
Add to Cart
Add to Cart

Voriconazole is a second-generation triazole, and is derived from fluconazole. It has a broad and enhanced antifungal spectrum as compared to other older triazoles. It is mostly well tolerated and is available in both oral and intravenous formulations.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 137234-62-9
Physical Form Neat
Linear Formula C16H14F3N5 O
MDL Number MFCD00905717
Synonym 2 R,3 S-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1 H-1,2,4-triazol-1-yl)butan-2-ol; UK-109496
Type Analytical Standard
Quantity 25 mg
Formula Weight 349.31
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.