Learn More
Med Vet International Vetrimycin 200 (Oxytetracycline Hydrochloride) Injection, Sterile Antibiotic, 100mL, Each
Supplier:  Med Vet International RXVETRI200100INJV
Encompass_Preferred
Vetrimycin 200 is a sterile, ready-to-use solution for the administration of the broad-spectrum antibiotic oxytetracycline by injection. Each mL contains 200 mg of oxytetracycline base as amphoteric oxytetracycline for use in beef cattle; dairy cattle; calves, including preruminating (veal) calves; and swine. Intended for use in treatment of the following diseases when due to oxytetracycline-susceptible organisms: CATTLE: Pneumonia and shipping fever complex associated with Pasteurella spp. and Hemophilus spp. ; infectious bovine kerato-conjunctivitis (pink eye) caused by Moraxella bovis; foot rot and diphtheria caused by Fusobacterium necrophorum; bacterial enteritis (scours) caused by Escherichia coli; wooden tongue caused by Actinobacillus lignieresii; leptospirosis caused by Leptospira pomona; and wound infections and acute metritis caused by strains of staphylococci and streptococci organisms sensitive to oxytetracycline.
Catalog No. 50-282-8428