Learn More
Med Vet International Vetrishield Plus Flavored Chewables For Dogs, Up To 25 Pounds, Blue Label, 6 Dose
Catalog No. 502827780
Encompass_Preferred
VetriShield Plus Flavored Chewables (ivermectin/pyrantel pamoate) are indicated for use in dogs to prevent canine heartworm disease by eliminating the tissue stage of heartworm larvae (Dirolaria immitis) for a month (30 days) after infection and for the treatment and control of ascarids (Toxocara canis, Toxascaris leonina) and hookworms (Ancylostoma caninum, Uncinaria stenocephala, Ancylostoma braziliense). For use in dogs six weeks of age and older.
Catalog No. 50-282-7780 Supplier Med Vet International Supplier No. RXVETRIS6V
VetriShield Plus Flavored Chewables (ivermectin/pyrantel pamoate) are indicated for use in dogs to prevent canine heartworm disease by eliminating the tissue stage of heartworm larvae (Dirolaria immitis) for a month (30 days) after infection and for the treatment and control of ascarids (Toxocara canis, Toxascaris leonina) and hookworms (Ancylostoma caninum, Uncinaria stenocephala, Ancylostoma braziliense). For use in dogs six weeks of age and older.