Learn More
Medchemexpress LLC Vevorisertib | 1416775-46-6 | 98.0% | C35H38N8O | 10MG
SDP
Catalog No. 5000259394
Encompass_Preferred
Vevorisertib is an orally active, selective pan-AKT serine/threonine kinase inhibitor provided for research use. It inhibits AKT1, AKT2, and AKT3 with low-nanomolar potency and is intended for in-vitro and preclinical studies; it is not for human use. The compound has formula C35H38N8O, molecular weight 586.73, and a reported purity of about 98%.
  • Potent inhibition of AKT isoforms at low nanomolar concentrations.
  • Suitable for in-vitro and preclinical cancer research.
  • High chemical purity appropriate for biological testing.
  • Well-characterized small-molecule with published identifiers and IUPAC name.
  • Available in small pack sizes to support laboratory workflows.
Catalog No. 50-002-59394 Supplier Medchemexpress LLC Supplier No. HY13745810MG
Vevorisertib is an orally active, selective pan-AKT serine/threonine kinase inhibitor provided for research use. It inhibits AKT1, AKT2, and AKT3 with low-nanomolar potency and is intended for in-vitro and preclinical studies; it is not for human use. The compound has formula C35H38N8O, molecular weight 586.73, and a reported purity of about 98%.
  • Potent inhibition of AKT isoforms at low nanomolar concentrations.
  • Suitable for in-vitro and preclinical cancer research.
  • High chemical purity appropriate for biological testing.
  • Well-characterized small-molecule with published identifiers and IUPAC name.
  • Available in small pack sizes to support laboratory workflows.