Learn More
Medchemexpress LLC Vevorisertib trihydrochloride | 1416775-08-0 | 99.13% | 696.11 | 25 MG
SDP
Catalog No. 5000351031
Encompass_Preferred
Vevorisertib trihydrochloride is a selective, allosteric, pan-AKT and AKT1-E17K mutant inhibitor. It potently inhibits phosphorylation of AKT with Kd values of 1.2 nM for AKT1 and 8.6 nM for AKT1-E17K, and IC50 values of 0.55, 0.81, and 1.3 nM for AKT1, AKT2, and AKT3, respectively. This compound can be used for cancer research.
  • Selective, allosteric, pan-AKT and AKT1-E17K mutant inhibitor
  • Potently inhibits phosphorylation of AKT
  • Used for cancer research
Catalog No. 50-003-51031 Supplier Medchemexpress LLC Supplier No. 25MGHY137458A
Vevorisertib trihydrochloride is a selective, allosteric, pan-AKT and AKT1-E17K mutant inhibitor. It potently inhibits phosphorylation of AKT with Kd values of 1.2 nM for AKT1 and 8.6 nM for AKT1-E17K, and IC50 values of 0.55, 0.81, and 1.3 nM for AKT1, AKT2, and AKT3, respectively. This compound can be used for cancer research.
  • Selective, allosteric, pan-AKT and AKT1-E17K mutant inhibitor
  • Potently inhibits phosphorylation of AKT
  • Used for cancer research