Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc vF 450 aMu Ly-6G Ly-6C Anti

Supplier:  Cytek Biosciences Inc 755931U100

Encompass_Preferred

Product Name: violetFluor 450 Anti-Mouse Ly-6G/Ly-6C (Gr-1) (RB6-8C5)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 450 Anti-Mouse Ly-6G/Ly-6C (Gr-1) (RB6-8C5)
Cytek Catalog Number: 75-5931-U100
Marker: Ly-6G/Ly-6C (Gr-1)
Target Species: Mouse
Clonality: Monoclonal
Clone: RB6-8C5
Host Species: Rat
Isotype: IgG2b, k
Conjugation: violetFluor 450
Size: 100 μg

Catalog No. 50-000-86671


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.