Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc vF 450 aMu TER-119 Anti

Catalog No. 5000086670
Encompass_Preferred

Product Name: violetFluor 450 Anti-Mouse TER-119 (TER-119)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 450 Anti-Mouse TER-119 (TER-119)
Cytek Catalog Number: 75-5921-U100
Marker: TER-119
Target Species: Mouse
Clonality: Monoclonal
Clone: TER-119
Host Species: Rat
Isotype: IgG2b, k
Conjugation: violetFluor 450
Size: 100 μg

Catalog No. 50-000-86670 Supplier Cytek Biosciences Inc Supplier No. 755921U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: violetFluor 450 Anti-Mouse TER-119 (TER-119)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 450 Anti-Mouse TER-119 (TER-119)
Cytek Catalog Number: 75-5921-U100
Marker: TER-119
Target Species: Mouse
Clonality: Monoclonal
Clone: TER-119
Host Species: Rat
Isotype: IgG2b, k
Conjugation: violetFluor 450
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.