Learn More
Biotium ViaFluor 488 Live Cell Microtubule Stain, Trial Size
Catalog No. NC1526410
Encompass_Preferred
ViaFluor™ 488 is a green fluorescent taxol probe with excitation/emission at 500/515 nm that allows fluorescent staining of microtubules in living cells for detection in the FITC channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.
Catalog No. NC1526410 Supplier Biotium Supplier No. 70062T
ViaFluor™ 488 is a green fluorescent taxol probe with excitation/emission at 500/515 nm that allows fluorescent staining of microtubules in living cells for detection in the FITC channel. Supplied at 1000X in DMSO, with a vial of 100 mM verapamil, which can enhance staining in some cell types.