Learn More
Felix Storch Inc ACCUCOLD Freezer 33.75'' H x 23.63'' W x 23.5'' D
SDP
Catalog No. NC1612700
Encompass_Preferred
Built-in undercounter -25C capable all-freezer with front control panel equipped with a digital thermostat and NIST calibrated thermometer/alarm 33.75' H x 23.63' W x 23.5' D: https://www.accucold.com/product/VT65MLBIMED2
Catalog No. NC1612700 Supplier Felix Storch Inc Supplier No. VT65MLBIMED2
Built-in undercounter -25C capable all-freezer with front control panel equipped with a digital thermostat and NIST calibrated thermometer/alarm 33.75' H x 23.63' W x 23.5' D: https://www.accucold.com/product/VT65MLBIMED2