Learn More
Aviva Systems Biology Corp VGF Antibody - middle region (ARP42128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VGF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VGF. Alternate names: -ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-58120