Learn More
Aviva Systems Biology Corp Vgll2 Antibody - N-terminal region (ARP50814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vgll2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C130057C21Rik, VITO-1, Vito1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-542