Learn More
Aviva Systems Biology Corp Vgll2 Antibody - C-terminal region (ARP37509_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37509_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vgll2. It was validated on Western Blot. Vgll2,vestigial like 2 homolog (Drosophila),Vito1; VITO-1; C130057C21Rik; Vgll2,transcription cofactor vestigial-like protein 2,transcription cofactor vestigial-like protein 2,vgl-2The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C130057C21Rik, VITO-1, Vito1ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-646