Learn More
Cayman Chemical VH 032 amide C7 Acid, Purity: ≥95%, CAS: 2172819-76-8, 25 mg
SDP
Catalog No. 502943066
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting ALK in cancer cells
Catalog No. 50-294-3066 Supplier Cayman Chemical Supplier No. 4044725 MG
A PROTAC precursor; has been used in the synthesis of PROTACs targeting ALK in cancer cells