Learn More
Cayman Chemical VH 032 LInker 1 100mg
SDP
Catalog No. 502935065
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting TEAD in cancer cells
Catalog No. 50-293-5065 Supplier Cayman Chemical Supplier No. 26148100 MG
A PROTAC precursor; has been used in the synthesis of PROTACs targeting TEAD in cancer cells