Learn More
Cayman Chemical VH 032 LInker 1 50mg
SDP
Catalog No. 502933928
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting TEAD in cancer cells
Catalog No. 50-293-3928 Supplier Cayman Chemical Supplier No. 2614850 MG
A PROTAC precursor; has been used in the synthesis of PROTACs targeting TEAD in cancer cells