Learn More
Cayman Chemical VH 032 Linker 2, Purity: ≥98%, CAS: 2064292-52-8, 25 mg
SDP
Catalog No. 502932788
Encompass_Preferred
A VHL Ligand 1 derivative; comprises the VHL ligand domain and PEG linker of the BET inhibitor-containing PROTACs MZ2 and MZP-55
Catalog No. 50-293-2788 Supplier Cayman Chemical Supplier No. 2614925 MG
A VHL Ligand 1 derivative; comprises the VHL ligand domain and PEG linker of the BET inhibitor-containing PROTACs MZ2 and MZP-55