Learn More
Cayman Chemical VH 032 LInker 2 50mg
SDP
Catalog No. 502934061
Encompass_Preferred
A VHL Ligand 1 derivative; comprises the VHL ligand domain and PEG linker of the BET inhibitor-containing PROTACs MZ2 and MZP-55
Catalog No. 50-293-4061 Supplier Cayman Chemical Supplier No. 2614950 MG
A VHL Ligand 1 derivative; comprises the VHL ligand domain and PEG linker of the BET inhibitor-containing PROTACs MZ2 and MZP-55