Learn More
Cayman Chemical VH 032 LInker 6 5mg
SDP
Catalog No. 502918702
Encompass_Preferred
A PROTAC precursor; has been used in the synthesis of PROTACs targeting Bcl-xL in senescent cells
Catalog No. 50-291-8702 Supplier Cayman Chemical Supplier No. 388985 MG
A PROTAC precursor; has been used in the synthesis of PROTACs targeting Bcl-xL in senescent cells