Learn More
Medchemexpress LLC VH 101, acid | 2408341-97-7 | 98.8% | 574.66 | C28H35FN4O6S | 10 MG
SDP
Catalog No. 5000299069
Encompass_Preferred
VH 101, acid is a functionalized von Hippel-Lindau (VHL) E3 ligase ligand developed for PROTAC research and conjugation chemistry. The molecule includes an E3 ligase recognition motif and an alkyl linker terminating in a primary amine, facilitating attachment to target protein ligands.
  • Designed for use as a VHL E3 ligase ligand in PROTAC development.
  • Alkyl linker with terminal amine enables straightforward conjugation chemistry.
  • High reported purity suitable for research applications.
  • Supplied in small research-scale quantities for synthetic work and assay testing.
Catalog No. 50-002-99069 Supplier Medchemexpress LLC Supplier No. HY4707010MG
VH 101, acid is a functionalized von Hippel-Lindau (VHL) E3 ligase ligand developed for PROTAC research and conjugation chemistry. The molecule includes an E3 ligase recognition motif and an alkyl linker terminating in a primary amine, facilitating attachment to target protein ligands.
  • Designed for use as a VHL E3 ligase ligand in PROTAC development.
  • Alkyl linker with terminal amine enables straightforward conjugation chemistry.
  • High reported purity suitable for research applications.
  • Supplied in small research-scale quantities for synthetic work and assay testing.