Learn More
Medchemexpress LLC VH 101, acid | 2408341-97-7 | 98.8% | C28H35FN4O6S | 100 MG
SDP
Catalog No. 5000359296
Encompass_Preferred
VH 101, acid is a functionalized von-Hippel-Lindau (VHL) protein ligand developed for PROTAC research and development. This compound contains an E3 ligase ligand and an alkyl linker with a terminal amine, making it suitable for conjugation to a target protein ligand. The VHL protein is a key substrate recognition subunit in Cullin RING E3 ubiquitin ligase complexes, often recruited by PROTACs to induce protein degradation.
  • Functionalized von-Hippel-Lindau (VHL) protein ligand
  • Designed for PROTAC research and development
  • Contains an E3 ligase ligand
  • Includes an alkyl linker with terminal amine
  • Ready for conjugation to target protein ligands
Catalog No. 50-003-59296 Supplier Medchemexpress LLC Supplier No. HY47070100MG
VH 101, acid is a functionalized von-Hippel-Lindau (VHL) protein ligand developed for PROTAC research and development. This compound contains an E3 ligase ligand and an alkyl linker with a terminal amine, making it suitable for conjugation to a target protein ligand. The VHL protein is a key substrate recognition subunit in Cullin RING E3 ubiquitin ligase complexes, often recruited by PROTACs to induce protein degradation.
  • Functionalized von-Hippel-Lindau (VHL) protein ligand
  • Designed for PROTAC research and development
  • Contains an E3 ligase ligand
  • Includes an alkyl linker with terminal amine
  • Ready for conjugation to target protein ligands