Learn More
Medchemexpress LLC VH 101, acid | 2408341-97-7 | 98.8% | C28H35FN4O6S | 25 MG
SDP
Catalog No. 5000358923
Encompass_Preferred
VH 101, acid is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC research and development. It contains an E3 ligase ligand plus an alkyl linker with a terminal amine, making it ready for conjugation to a target protein ligand.
  • Functionalized von-Hippel-Lindau (VHL) protein ligand
  • For PROTAC research and development
  • Contains an E3 ligase ligand
  • Alkyl linker with terminal amine for conjugation
  • VHL protein is a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes
  • Used by bifunctional Proteolysis-targeting chimeras (PROTACs) to induce ubiquitination and subsequent proteasomal degradation of a target protein
Catalog No. 50-003-58923 Supplier Medchemexpress LLC Supplier No. 25MGHY47070
VH 101, acid is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC research and development. It contains an E3 ligase ligand plus an alkyl linker with a terminal amine, making it ready for conjugation to a target protein ligand.
  • Functionalized von-Hippel-Lindau (VHL) protein ligand
  • For PROTAC research and development
  • Contains an E3 ligase ligand
  • Alkyl linker with terminal amine for conjugation
  • VHL protein is a substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes
  • Used by bifunctional Proteolysis-targeting chimeras (PROTACs) to induce ubiquitination and subsequent proteasomal degradation of a target protein