Learn More
Medchemexpress LLC VH 101 phenol-alkylC4-amine dihydrochloride | 2564467-03-2 | 676.67 | 1MG
SDP
Catalog No. 5000352413
Encompass_Preferred
VH 101 phenol-alkylC4-amine dihydrochloride is a von-Hippel-Lindau protein ligand (VHL), which incorporates an E3 ligase ligand and an alkylC4 linker. It can be utilized for PROTAC research.
  • Von-Hippel-Lindau protein ligand (VHL)
  • Incorporates an E3 ligase ligand
  • Incorporates an alkylC4 linker
  • Suitable for PROTAC research
  • Extracted from patent WO2022051326A1
Catalog No. 50-003-52413 Supplier Medchemexpress LLC Supplier No. HY1393351MG
VH 101 phenol-alkylC4-amine dihydrochloride is a von-Hippel-Lindau protein ligand (VHL), which incorporates an E3 ligase ligand and an alkylC4 linker. It can be utilized for PROTAC research.
  • Von-Hippel-Lindau protein ligand (VHL)
  • Incorporates an E3 ligase ligand
  • Incorporates an alkylC4 linker
  • Suitable for PROTAC research
  • Extracted from patent WO2022051326A1