Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH 101 phenol-alkylC4-amine dihydrochloride | 2564467-03-2 | >95.0% | C30H44Cl2FN5O5S | 10MG
SDP

Catalog No. 5000261083
Encompass_Preferred

VH 101 phenol-alkylC4-amine dihydrochloride is a von Hippel-Lindau (VHL) protein ligand developed as a building block for PROTAC synthesis and E3 ligase conjugation. Supplied as the dihydrochloride salt, it is intended for research use in targeted protein degradation and conjugation workflows.

  • VHL ligand suitable for PROTAC synthesis and E3 ligase recruitment.
  • Dihydrochloride salt for improved handling and conjugation chemistry.
  • Molecular formula C30H44Cl2FN5O5S; molecular weight 676.67 g/mol.
  • Purity reported as ≥95.0% by HPLC in supplier documentation.
  • Recommended storage: keep dry at room temperature; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Intended for research use in targeted protein degradation and conjugation workflows.

Catalog No. 50-002-61083 Supplier Medchemexpress LLC Supplier No. HY13933510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH 101 phenol-alkylC4-amine dihydrochloride is a von Hippel-Lindau (VHL) protein ligand developed as a building block for PROTAC synthesis and E3 ligase conjugation. Supplied as the dihydrochloride salt, it is intended for research use in targeted protein degradation and conjugation workflows.

  • VHL ligand suitable for PROTAC synthesis and E3 ligase recruitment.
  • Dihydrochloride salt for improved handling and conjugation chemistry.
  • Molecular formula C30H44Cl2FN5O5S; molecular weight 676.67 g/mol.
  • Purity reported as ≥95.0% by HPLC in supplier documentation.
  • Recommended storage: keep dry at room temperature; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Intended for research use in targeted protein degradation and conjugation workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.