Learn More
Medchemexpress LLC VH 101 phenol-alkylC6-amine dihydrochloride | 2564467-16-7 | 704.72 | 100 MG
SDP
Catalog No. 5000351320
Encompass_Preferred
VH 101 phenol-alkylC6-amine dihydrochloride is a functionalized von-Hippel-Lindau (VHL) protein-ligand that can be used for PROTAC synthesis.
  • Functionalized von-Hippel-Lindau (VHL) protein-ligand
  • Can be used for PROTAC synthesis
Catalog No. 50-003-51320 Supplier Medchemexpress LLC Supplier No. HY139334100MG
VH 101 phenol-alkylC6-amine dihydrochloride is a functionalized von-Hippel-Lindau (VHL) protein-ligand that can be used for PROTAC synthesis.
  • Functionalized von-Hippel-Lindau (VHL) protein-ligand
  • Can be used for PROTAC synthesis