Learn More
Medchemexpress LLC Vh 101 phenol-alkylC6-amine dihydrochloride | 2564467-16-7 | ≥95% | 704.72 | 250 MG
SDP
Catalog No. 5000351137
Encompass_Preferred
VH 101 phenol-alkylC6-amine dihydrochloride is a functionalized von-Hippel-Lindau (VHL) protein-ligand designed for optimal PROTAC (Proteolysis Targeting Chimeras) functionality. It binds the VHL protein, enabling ubiquitination of target proteins, and the terminal amine on its C6 linker allows conjugation to target ligands for PROTAC synthesis.
  • Functionalized VHL protein-ligand.
  • Designed for optimal PROTAC functionality.
  • Binds VHL protein to recruit E3 ligase.
  • Enables ubiquitination of target proteins.
  • Terminal amine on C6 linker allows conjugation to target ligands.
  • Used in PROTAC synthesis against various targets.
Catalog No. 50-003-51137 Supplier Medchemexpress LLC Supplier No. 250MGHY139334
VH 101 phenol-alkylC6-amine dihydrochloride is a functionalized von-Hippel-Lindau (VHL) protein-ligand designed for optimal PROTAC (Proteolysis Targeting Chimeras) functionality. It binds the VHL protein, enabling ubiquitination of target proteins, and the terminal amine on its C6 linker allows conjugation to target ligands for PROTAC synthesis.
  • Functionalized VHL protein-ligand.
  • Designed for optimal PROTAC functionality.
  • Binds VHL protein to recruit E3 ligase.
  • Enables ubiquitination of target proteins.
  • Terminal amine on C6 linker allows conjugation to target ligands.
  • Used in PROTAC synthesis against various targets.