Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH 101 thiol | 2358775-81-0 | 98.7% | 534.67 g·mol⁻¹ | C25H31FN4O4S2 | 10 MG
SDP

Catalog No. 5000299235
Encompass_Preferred

Thiol-functionalized VHL ligand used as a recruiting moiety for von Hippel-Lindau (VHL) E3 ligase in the synthesis of PROTACs. Supplied as a high-purity powder suitable for conjugation and synthetic chemistry applications, with recommended cold storage to preserve stability.

  • Thiol-functionalized VHL ligand for PROTAC synthesis
  • Molecular formula C25H31FN4O4S2; molecular weight 534.67 g·mol⁻¹
  • High purity (98.66%) suitable for synthesis and conjugation
  • Recommended storage: powder at -20°C (stable up to 3 years)
  • Pack size: 10 mg (other sizes available)
  • CAS number 2358775-81-0 for unambiguous identification

Catalog No. 50-002-99235 Supplier Medchemexpress LLC Supplier No. HY4785110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Thiol-functionalized VHL ligand used as a recruiting moiety for von Hippel-Lindau (VHL) E3 ligase in the synthesis of PROTACs. Supplied as a high-purity powder suitable for conjugation and synthetic chemistry applications, with recommended cold storage to preserve stability.

  • Thiol-functionalized VHL ligand for PROTAC synthesis
  • Molecular formula C25H31FN4O4S2; molecular weight 534.67 g·mol⁻¹
  • High purity (98.66%) suitable for synthesis and conjugation
  • Recommended storage: powder at -20°C (stable up to 3 years)
  • Pack size: 10 mg (other sizes available)
  • CAS number 2358775-81-0 for unambiguous identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.