Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VH 101, thiol | 2358775-81-0 | 98.7% | C25H31FN4O4S2 | 100 MG
SDP

Catalog No. 5000358299
Encompass_Preferred

VH 101, thiol (compound 20) is a VHL ligand used for the synthesis of PROTACs.

  • Acts as a VHL ligand for the synthesis of PROTACs.
  • Appears as a solid.
  • Soluble in DMSO (100 mg/mL).
  • Applicable in cancer and cancer targeted therapy research.
  • Part of the PROTAC ligands for E3 ligase category.
  • Can be found in the ubiquitination compound library.
  • Inhibits E3 ligase-recruiting moiety.
  • Target for biological activity is VHL.
  • Molecular weight is 534.67.

Catalog No. 50-003-58299 Supplier Medchemexpress LLC Supplier No. HY47851100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VH 101, thiol (compound 20) is a VHL ligand used for the synthesis of PROTACs.

  • Acts as a VHL ligand for the synthesis of PROTACs.
  • Appears as a solid.
  • Soluble in DMSO (100 mg/mL).
  • Applicable in cancer and cancer targeted therapy research.
  • Part of the PROTAC ligands for E3 ligase category.
  • Can be found in the ubiquitination compound library.
  • Inhibits E3 ligase-recruiting moiety.
  • Target for biological activity is VHL.
  • Molecular weight is 534.67.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.